Merck
  • Determination of rotenone in river water utilizing packed capillary column switching liquid chromatography with UV and time-of-flight mass spectrometric detection
Determination of rotenone in river water utilizing packed capillary column switching liquid chromatography with UV and time-of-flight mass spectrometric detection
Journal of Chromatography A (2003)
Holm A, et al.