Merck
  • Analysis of major components of residual bacitracin and colistin in food samples by liquid chromatography tandem mass spectrometry
Analysis of major components of residual bacitracin and colistin in food samples by liquid chromatography tandem mass spectrometry
Analytica Chimica Acta (2005)
Sin DW, et al.