Merck
  • Disruption of angiogenesis by PEX, a noncatalytic metalloproteinase fragment with integrin binding activity.
Disruption of angiogenesis by PEX, a noncatalytic metalloproteinase fragment with integrin binding activity.
Cell (1998-02-26)
P C Brooks, S Silletti, T L von Schalscha, M Friedlander, D A Cheresh
ZUSAMMENFASSUNG
Angiogenesis depends on both cell adhesion and proteolytic mechanisms. In fact, matrix metalloproteinase 2 (MMP-2) and integrin alphavbeta3 are functionally associated on the surface of angiogenic blood vessels. A fragment of MMP-2, which comprises the C-terminal hemopexin-like domain, termed PEX, prevents this enzyme binding to alphavbeta3 and blocks cell surface collagenolytic activity. PEX blocks MMP-2 activity on the chick chorioallantoic membrane where it disrupts angiogenesis and tumor growth. Importantly, a naturally occurring form of PEX can be detected in vivo in conjunction with alphavbeta3 expression in tumors and during developmental retinal neovascularization. Levels of PEX in these vascularized tissues suggest that it interacts with endothelial cell alphavbeta3 where it serves as a natural inhibitor of MMP-2 activity, thereby regulating the invasive behavior of new blood vessels.