Merck
  • Single-isomer carboxymethyl-γ-cyclodextrin as chiral resolving agent for capillary electrophoresis
Single-isomer carboxymethyl-γ-cyclodextrin as chiral resolving agent for capillary electrophoresis
Journal of Chromatography A (2016)
Benkovics G, et al.