Merck
  • Cucurbituril and α-and β-Cyclodextrins as Ligands for the Complexation of Nonionic Surfactants and Polyethyleneglycols in Aqueous Solutions
Cucurbituril and α-and β-Cyclodextrins as Ligands for the Complexation of Nonionic Surfactants and Polyethyleneglycols in Aqueous Solutions
Journal of Inclusion Phenomena and Macrocyclic Chemistry (2000)
Buschmann HJ, et al.