Merck
  • HPLC-fluorescence determination of amino acids in pharmaceuticals after pre-column derivatization with phanquinone
HPLC-fluorescence determination of amino acids in pharmaceuticals after pre-column derivatization with phanquinone
Journal of Pharmaceutical and Biomedical Analysis (2004)
Gatti.R, et al.