Merck
  • RP-HPLC method development and validation for simultaneous estimation of lopinavir and ritonavir in dosage form and in plasma
RP-HPLC method development and validation for simultaneous estimation of lopinavir and ritonavir in dosage form and in plasma
International Journal of Pharmaceutical Sciences Review and Research (2014)
Kumar VK, et al.