Merck
  • Separation and determination of pharmaceutically important polyols in dosage forms by capillary isotachophoresis
Separation and determination of pharmaceutically important polyols in dosage forms by capillary isotachophoresis
Journal of Chromatography A (1997)
Pospisilova M, et al.