Merck
  • Prohibitin is expressed in pancreatic beta-cells and protects against oxidative and proapoptotic effects of ethanol.
Prohibitin is expressed in pancreatic beta-cells and protects against oxidative and proapoptotic effects of ethanol.
The FEBS journal (2009-12-25)
Jong Han Lee, K Hoa Nguyen, Suresh Mishra, B L Grégoire Nyomba
ZUSAMMENFASSUNG
Pancreatic beta-cell dysfunction is a prerequisite for the development of type 2 diabetes. Alcoholism is a diabetes risk factor and ethanol increases oxidative stress in beta-cells, whereas the mitochondrial chaperone prohibitin (PHB) has antioxidant effects in several cell types. In the present study we investigated whether PHB is expressed in beta-cells and protects these cells against deleterious effects of ethanol, using INS-1E and RINm5F beta-cell lines. Endogenous PHB was detected by western blot and immunocytochemistry. Reactive oxygen species were determined by 5-(and-6)-chloromethyl-2',7'-dichlorodihydrofluorescein diacetate fluorescence assay, and mitochondrial activity was assessed by 3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyl-tetrazolium bromide (MTT) reduction, uncoupling protein 2 expression and ATP production. Cell death was determined by Hoechst 33342 staining, cleaved caspase-3 levels and flow cytometry. PHB was expressed in beta-cells under normal conditions and colocalized with Hoechst 33342 in the nucleus and with the mitochondrial probe Mitofluor in the perinuclear area. In ethanol-treated cells, MTT reduction and ATP production decreased, whereas reactive oxygen species, uncoupling protein 2 and cleaved caspase-3 levels increased. In addition, flow cytometry analysis showed an increase of apoptotic cells. Ethanol treatment increased PHB expression and induced PHB translocation from the nucleus to the mitochondria. PHB overexpression decreased the apoptotic effects of ethanol, whereas PHB knockdown enhanced these effects. The protective effects of endogenous PHB were recapitulated by incubation of the cells with recombinant human PHB. Thus, PHB is expressed in beta-cells, increases with oxidative stress and protects the cells against deleterious effects of ethanol.