Merck
  • B Vitamins in the nervous system: Current knowledge of the biochemical modes of action and synergies of thiamine, pyridoxine, and cobalamin
B Vitamins in the nervous system: Current knowledge of the biochemical modes of action and synergies of thiamine, pyridoxine, and cobalamin
CNS Neuroscience & Therapeutics (2019)
Calderon-Ospina CA, et al.