Merck
  • Iron-binding proteins in sputum of chronic bronchitis patients with Haemophilus influenzae infections
Iron-binding proteins in sputum of chronic bronchitis patients with Haemophilus influenzae infections
The European Respiratory Journal (1997)
Vogel L, et al.