Merck
  • A Rapid and Green Method for the Determination of Veterinary Pharmaceuticals in Swine Wastewater by Fluorescence Spectrophotometry.
A Rapid and Green Method for the Determination of Veterinary Pharmaceuticals in Swine Wastewater by Fluorescence Spectrophotometry.
Bulletin of environmental contamination and toxicology (2019-08-31)
Karina León-Aguirre, Emanuel Hernández-Núñez, Avel González-Sánchez, Roger Méndez-Novelo, Carmen Ponce-Caballero, Germán Giácoman-Vallejos
ZUSAMMENFASSUNG
A growing concern exists over water contamination by veterinary pharmaceuticals from small pig farms in Yucatan, Mexico, where the anaerobic digesters installed as the wastewater treatment system are not operated properly. Therefore, considerable interest exists to develop analytical methods to detect these compounds and characterize their fate in the environment. In this study, the detection of three antibiotics (enrofloxacin, oxytetracycline and sulfamethoxazole) and a β-agonist (ractopamine) was carried out using fluorescence spectrophotometry, with a semi-quantitative approach and a low environmental impact. Wastewater samples from 10 pig farms were analyzed, detecting concentrations of approximately 0.043 μg mL-1 for enrofloxacin, 1.427 μg mL-1 for oxytetracycline, and 9.748 μg mL-1 for sulfamethoxazole. The detection of these pharmaceuticals in the effluents of treated wastewater from the biodigesters of the pig farms suggests the need to optimize the system and prevent the entry of these compounds into the environment.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Enrofloxacin, VETRANAL®, analytical standard