Merck
  • The effect of chronic oral vitamin D supplementation on adiposity and insulin secretion in hypothalamic obese rats.
The effect of chronic oral vitamin D supplementation on adiposity and insulin secretion in hypothalamic obese rats.
The British journal of nutrition (2019-03-30)
Zoé M Guareschi, Ana C Valcanaia, Vanessa M Ceglarek, Pamela Hotz, Bruna K Amaral, Domwesley W de Souza, Thainan A de Souza, Tarlliza Nardelli, Thiago R Ferreira, Nayara C Leite, Camila Lubackzeuski, Henriette R de O Emilio, Sabrina Grassiolli
ZUSAMMENFASSUNG
Reduced plasma vitamin D (VD) levels may contribute to excessive white adipose tissue, insulin resistance (IR) and dyslipidaemia. We evaluated the effect of chronic oral VD supplementation on adiposity and insulin secretion in monosodium glutamate (MSG)-treated rats. During their first 5 d of life, male neonate rats received subcutaneous injections of MSG (4 g/kg), while the control (CON) group received saline solution. After weaning, groups were randomly distributed into VD supplemented (12 µg/kg; three times/week) and non-supplemented (NS) rats, forming four experimental groups (n 15 rats/group): CON-NS, CON-VD, MSG-NS and MSG-VD. At 76 d of life, rats were submitted to an oral glucose tolerance test (OGTT; 2 g/kg), and at 86 d, obesity, IR and plasma metabolic parameters were evaluated. Pancreatic islets were isolated for glucose-induced insulin secretion (GIIS), cholinergic insulinotropic response and muscarinic 3 receptor (M3R), protein kinase C (PKC) and protein kinase A (PKA) expressions. Pancreas was submitted to histological analyses. VD supplementation decreased hyperinsulinaemia (86 %), hypertriacylglycerolaemia (50 %) and restored insulin sensibility (89 %) in MSG-VD rats, without modifying adiposity, OGTT or GIIS, compared with the MSG-NS group. The cholinergic action was reduced (57 %) in islets from MSG-VD rats, without any change in M3R, PKA or PKC expression. In conclusion, chronic oral VD supplementation of MSG-obese rats was able to prevent hyperinsulinaemia and IR, improving triacylglycerolaemia without modifying adiposity. A reduced cholinergic pancreatic effect, in response to VD, could be involved in the normalisation of plasma insulin levels, an event that appears to be independent of M3R and its downstream pathways.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-α-Tubulin-Antikörper, monoklonaler Antikörper der Maus gegen, clone DM1A, purified from hybridoma cell culture