Merck
  • Chronic activation of Mas-related gene receptors (Mrg) reduces the potency of morphine-induced analgesia via PKC pathway in naive rats.
Chronic activation of Mas-related gene receptors (Mrg) reduces the potency of morphine-induced analgesia via PKC pathway in naive rats.
Brain research (2019-08-09)
Zhijing Hu, Tian Zhao, Tingjun Chen, Dongmei Wang
ZUSAMMENFASSUNG
Mas oncogene-related gene receptors (Mrg) are uniquely distributed in small and medium cells of trigeminal and dorsal root ganglia (DRG). The physiological and pharmacological properties of Mrg are unknown. We have shown that intermittent activation of MrgC prevents and reverses morphine tolerance. Now we observed that intrathecal (i.t.) administration of the MrgC agonist bovine adrenal medulla 8-22 (BAM8-22, 3 nmol) for 3 and 6 days reduced the potency of morphine analgesia by 1.5 and 3.5 folds, respectively. Daily administration of BAM8-22 for 6 days also significantly decreased the tail flick latency. The administration of another MrgC agonist (Tyr6)-γ2-MSH-6-12 (MSH, 3 nmol) reduced morphine potency and the reduction was abolished following the co-administration of the protein kinase C (PKC) inhibitor chelerythrine chloride (CLT, 3 nmol). The chronic treatment with BAM8-22 or MSH increased the expression of PKC-gamma (PKCγ) in the cell membrane of spinal dorsal horn neurons and PKC-epsilon (PKCε) in the cell membrane and cytosol of DRG neurons. Moreover, the BAM8-22 treatment induced an increase in the expression of calcitonin gene-related peptide (CGRP) and neuronal nitric oxide synthase (nNOS) in small and medium cells in DRG. All of these responses were not seen when BAM8-22 or MSH was co-administered with the PKC inhibitor CLT (3 nmol) or GF-109203X (10 nmol). The present study suggested that the chronic activation of MrgC upregulated expressions of pronociceptive mediators via PKC signaling pathway leading to the suppression of antinociceptive property of morphine. These effects are opposite to those occurred when MrgC is activated acutely or moderately.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Chelerythrinchlorid, ≥95% (TLC), powder
Sigma-Aldrich
GF 109203X, synthetic, ≥90% (HPLC)