Merck
  • Protective effect of Jie-Geng-Tang against Staphylococcus aureus induced acute lung injury in mice and discovery of its effective constituents.
Protective effect of Jie-Geng-Tang against Staphylococcus aureus induced acute lung injury in mice and discovery of its effective constituents.
Journal of ethnopharmacology (2019-07-12)
Yining Liu, Zhuping Hong, Jing Qian, Yi Wang, Shufang Wang
ZUSAMMENFASSUNG
Jie-Geng-Tang (JGT), a famous traditional Chinese medicine prescription, consists of Platycodonis Radix and Glycyrrhizae Radix et Rhizoma. According to traditional medicinal theory, JGT exerts various effects, including apocenosis, detoxifying, moisturizing the lung and relieving sore throat. It is often used to treat throat inflammation and lung diseases. To determine the protective effect of JGT on Staphylococcus aureus (S. aureus)-induced acute lung injury (ALI) in mice and to identify the compounds in the prescription that may be responsible for antibacterial activity. The protective effect of JGT was assessed using S. aureus-induced ALI mice (i.g., 2.7 g/kg/day). Bacterial burden, pathological morphology, cytokine levels of TNF-α, IL-1β, KC, and MIP-2 were evaluated in the lung and bronchoalveolar lavage fluid at 24 h post-infection, respectively. Twenty three compounds in the prescription were evaluated for their minimum inhibitory concentration (MIC) in vitro by means of microbroth dilution method against S. aureus. The antibacterial effects in vitro of licochalcone A and isoliquiritigenin were also investigated by transmission electron microscopy. In vivo antibacterial activities of licochalcone A and isoliquiritigenin were evaluated by survival rates, bacterial burden, and pathological morphology of lung tissues on S. aureus-induced ALI in mice (i.p., 160 mg/kg/day). Pretreatment with JGT significantly improved the pathological morphology of lung tissues on S. aureus-induced ALI in mice, accompanied with the reduced bacterial burden in the lungs and inhibited expression of inflammatory cytokine levels at 24 h post-infection. Five compounds, namely licochalcone A, licoisoflavone B, glyasperin A, isoliquiritigenin, and licochalcone B from Jie-Geng-Tang displayed good antibacterial activities against S. aureus (MIC < 128 μg/mL). Furthermore, applications of licochalcone A and isoliquiritigenin resulted in the increased survival rates, reduced bacterial burden in the lungs, and improved pathological morphology of lung tissues in S. aureus infected mice. The study demonstrated that Jie-Geng-Tang presented protective role of acute lung injury, which supported its traditional use for the treatment of lung diseases. Licochalcone A, isoliquiritigenin, licoisoflavone B, glyasperin A, and licochalcone B might contribute to the antibacterial activity of JGT on S. aureus-induced acute lung injury. The anti-S. aureus activity of licoisoflavone B, glyasperin A, and licochalcone B in vitro, as well as the anti-S. aureus activity of licochalcone A in vivo, were first reported in this study.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Licochalcon A, ≥96.0% (HPLC)