Merck
Exogenous expression of caveolin-1 is sufficient for hepatic stellate cell activation.
Journal of cellular biochemistry (2019-07-02)
Mariana Ilha, Ketlen da Silveira Moraes, Francieli Rohden, Leo Anderson Meira Martins, Radovan Borojevic, Guido Lenz, Florencia Barbé-Tuana, Fátima Costa Rodrigues Guma
ZUSAMMENFASSUNG
Caveolin-1 (Cav-1) expression is increased in hepatic stellate cells (HSC) upon liver cirrhosis and it functions as an integral membrane protein of lipid rafts and caveolae that regulates and integrates multiple signals as a platform. This study aimed to evaluate the role of Cav-1 in HSC. Thus, the effects of exogenous expression of Cav-1 in GRX cells, a model of activated HSC, were determined. Here, we demonstrated through evaluating well-known HSC activation markers - such as α-smooth muscle actin, collagen I, and glial fibrillary acidic protein - that up regulation of Cav-1 induced GRX to a more activated phenotype. GRXEGFP-Cav1 presented an increased migration, an altered adhesion pattern, a reorganization f-actin cytoskeleton, an arrested cell cycle, a modified cellular ultrastructure, and a raised endocytic flux. Based on this, GRX EGFP-Cav1 represents a new cellular model that can be an important tool for understanding of events related to HSC activation. Furthermore, our results reinforce the role of Cav-1 as a molecular marker of HSC activation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Osmiumtetroxid, ReagentPlus®, 99.8%
Sigma-Aldrich
Toluidinblau O, Technical grade
Sigma-Aldrich
Toluidinblau, suitable for microscopy, 8.74% (ZN (THEORY))
Sigma-Aldrich
Sulforhodamin B Mononatriumsalz, Dye content 75 %