Merck
  • miR-107 regulates growth and metastasis of gastric cancer cells via activation of the PI3K-AKT signaling pathway by down-regulating FAT4.
miR-107 regulates growth and metastasis of gastric cancer cells via activation of the PI3K-AKT signaling pathway by down-regulating FAT4.
Cancer medicine (2019-07-13)
Li Wang, Kunkun Li, Chen Wang, Xiaoxin Shi, Huiyu Yang
ZUSAMMENFASSUNG
To investigate the effect of miR-107 on the growth and metastasis of gastric cancer (GC) and elucidate the probable mechanisms. The expression of miR-107 and FAT4 in GC tissues and cells were detected using qRT-PCR. Bioinformatics and dual luciferase reporter gene assays were used to analyze the relationship between miR-107 and FAT4. miR-NC, miR-107 inhibitor, pcDNA3.1-FAT4 and siRNA-FAT4 were transfected into AGS and MKN-45 GC cell lines, respectively. The proliferation and migration abilities of GC cells after transfection were evaluated using the MTT assay, scratch test and transwell assay. The expression of epithelial-mesenchymal transition (EMT) markers: E-cadherin, N-cadherin, vimentin and related proteins of the PI3K/AKT signaling pathway were determined using western blot. The xenograft tumors of nude mice were observed to assess the tumorigenicity of GC cells in vivo. MiR-107 was up-regulated, while FAT4 was down-regulated in GC tissues and cells (P < 0.05); FAT4 was targeted and negatively regulated by miR-107. Down-regulating miR-107 or up-regulating FAT4 inhibited the GC cells proliferation, migration, invasion and tumorigenicity, and could also reduce the expression of N-cadherin, vimentin, p-PI3K and p-Akt expression and up-regulate E-cadherin. miR-107 promotes growth and metastasis in GC via activation of PI3K-AKT signaling by targeting FAT4, which may be a target for GC treatment.