Merck
Partial (two-thirds) nephrectomy in pigs: A comparison of three surgical approaches.
Research in veterinary science (2017-11-14)
Hai-Feng Liu, Wei-Jing Meng, Xiang-Dong Kong, Ge Bai, Hui Li, Jian-Tao Zhang, Hong-Gang Fan, Hong-Bin Wang
ZUSAMMENFASSUNG
Partial (2/3) nephrectomy can be performed via the laparoscopic, retroperitoneal, and transperitoneal approach. Outcomes of the three approaches were compared in this study. 2/3 nephrectomy were performed in 21 healthy Bama miniature pigs (mean bodyweight 20.59±2.78kg). Pigs were divided into three groups: those that underwent 2/3 nephrectomy via laparoscopy (LN group, n=7), the retroperitoneal approach (RN group, n=7), or the transperitoneal approach (TN group, n=7). We monitored pre- and postoperative physiologic parameters, blood cell count, and stress and renal function biomarkers. Differences among groups were analyzed. 2/3 nephrectomy was successfully performed in all pigs without any complications. Mean surgical time in the LN group (60.71±7.34min) and the TN group (58.57±4.72min) was significantly longer than that in the RN group (41.14±5.33min). Warm ischemia in the LN group (38±7.57min) was significantly longer than that in the TN group (28.86±4.53min), which was significantly longer than that in the RN group (17.86±2.34min). The postoperative serum concentration of C-reactive protein in the TN group was significantly higher than that in the LN group (p<0.05). So retroperitoneal approach was best choice in case of bilateral renal lesion resulted in shortest ischemia time, and laparoscopic partial nephrectomy should be the primary choice in majority situations resulted in less body stress, smaller surgical incisions and less blood loss.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cefazolin Natriumsalz, 89.1-110.1%