Merck
  • Interleukin-6 and tumour necrosis factor-α cooperatively promote cell cycle regulators and proliferate rheumatoid arthritis fibroblast-like synovial cells.
Interleukin-6 and tumour necrosis factor-α cooperatively promote cell cycle regulators and proliferate rheumatoid arthritis fibroblast-like synovial cells.
Scandinavian journal of rheumatology (2019-10-22)
K Kaneshiro, Y Sakai, K Suzuki, K Uchida, K Tateishi, Y Terashima, Y Kawasaki, N Shibanuma, K Yoshida, A Hashiramoto
ZUSAMMENFASSUNG
Objective: To elucidate the roles of interleukin-6 (IL-6) and tumour necrosis factor-α (TNF-α) in cell cycle regulation and proliferation of rheumatoid arthritis fibroblast-like synovial cells (RA-FLSs). Methods: Under stimulation with IL-6/soluble interleukin-6 receptor (sIL-6R) and TNF-α, we examined the expression of cell cycle regulators [p16INK4a, p21Cip1, p27Kip1, cyclin-dependent kinase-4 (CDK4), CDK6, Cyclin D, Cyclin E, and retinoblastoma protein (pRB)] by quantitative polymerase chain reaction, Western blotting, and immunofluorescence staining. The expression of pRB, with or without 10% foetal bovine serum, was examined by Western blotting. DNA synthesis and cell viability were examined by the BrdU assay and WST-8 assay, respectively. After transfection with siRNA/p16INK4a, siRNA/p21Cip1, siRNA/p27Kip1, siRNA/CDK4, or siRNA/CDK6, RA-FLSs were successively stimulated with or without IL-6/sIL-6R or TNF-α to determine cell viability. Results: IL-6/sIL-6R significantly decreased the expression of p16INK4a, and increased p21Cip1, Cyclin E1, CYCLIN D, and pRB. TNF-α decreased the expression of CDK4, and significantly increased p27Kip1, CDK6, Cyclin E1/E2, CYCLIN D, CYCLIN E, pRB, and phosphorylated pRB (phospho-pRB). By immunofluorescence staining, CYCLIN D and phospho-pRB were simultaneously stained in the single cell. In serum-free culture, the expression of pRB was apparently decreased. DNA synthesis and cell viability were significantly increased by IL-6/sIL-6R and TNF-α. Silencing of CDK6 attenuated the cell viability induced by IL-6 and TNF-α. Conclusion: The results indicate that IL-6 and TNF-α interact with each other in regulating the cell cycle and accelerate the proliferation of RA-FLSs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin-Fragment 26-33 Amid, ≥97% (HPLC), powder