Merck
  • Enhanced adsorption of steroid estrogens by one-pot synthesized phenyl-modified mesoporous silica: Dependence on phenyl-organosilane precursors and pH condition.
Enhanced adsorption of steroid estrogens by one-pot synthesized phenyl-modified mesoporous silica: Dependence on phenyl-organosilane precursors and pH condition.
Chemosphere (2019-06-23)
Pei Gao, Zhijie Liang, Zhiwei Zhao, Wenhao Wang, Chun Yang, Bibo Hu, Fuyi Cui
ZUSAMMENFASSUNG
In this study, the phenyl-modified mesoporous materials were successfully synthesized using phenyl-organosilanes (trimethoxyphenylsilane and triethoxyphenylsilanea) by one-pot co-condensation method for the removal of estrone (E1), 17β-estradiol (E2), and 17α-ethinyl estradiol (EE2). Both the triethoxyphenylsilane-modified material (20%EtPh-MCM-41) and trimethoxyphenylsilane-modified material (20%MePh-MCM-41) could rapidly achieve equilibrium in 30 min at low adsorbent dosage of 0.025 g L-1. But the different hydrolysable groups of trimethoxyphenylsilane and triethoxyphenylsilane led to the discrepancies in physicochemical properties of the 20%EtPh-MCM-41 and 20%MePh-MCM-41, and thus affected adsorption performance. The 20%EtPh-MCM-41 exhibited the faster estrogen adsorption rates expressed in pseudo-second-order kinetic constant than the 20%MePh-MCM-41 due to the more hydrophobicity. Conversely, the 20%MePh-MCM-41 had much more estrogen adsorption capacities than the 20%EtPh-MCM-41 because of the more available adsorption sites. The addition of the phenyl-organosilane improved estrogen adsorption by π-π and hydrophobic interactions, and the Langmuir-model-based maximum adsorption amounts could reach 99.02, 83.47, and 53.60 mg g-1 for EE2, E2, and E1, respectively. But excessive concentration of phenyl-organosilane decreased adsorption capacities due to poor pore structure. Alkaline solution, which induced estrogen deprotonation and negative surface charge of absorbents, inhibited estrogen adsorption by electrostatic repulsion and the decreased hydrophobic interaction, but acidic and neutral solutions, ionic strength, and humic acid did not significantly affect estrogen removal. This work not only showed the high potential of trimethoxyphenylsilane-modified MCM-41 used in water purification for steroid estrogens, but also demonstrated the suitable selection of organosilane precursors was key in producing favorable materials with designed functionality.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Estron, ≥99%