Merck
  • DKK1 expression is suppressed by miR-9 during induced dopaminergic differentiation of human trabecular meshwork mesenchymal stem cells.
DKK1 expression is suppressed by miR-9 during induced dopaminergic differentiation of human trabecular meshwork mesenchymal stem cells.
Neuroscience letters (2019-06-08)
Elham Sadat Mirfazeli, Ehsan Arefian, Samad Nadri, Rezvan Rezazadeh Valojerdi, Mousa Kehtari, Bahman Zeynali
ZUSAMMENFASSUNG
Wnt/β-catenin pathway has been recently identified as one of the key players in dopaminergic (DA) neuron differentiation. DKK1, the potent inhibitor of the Wnt/β-catenin pathway, is expressed in a precisely controlled manner in ventral midbrain during brain development, however the molecular mechanism underlying this regulation is still unknown. Here we show that human trabecular meshwork mesenchymal stem cells (TM-MSCs) can be used as an efficient tool for in vitro differentiation of DA neurons. After differentiating TM-MSCs to DA neuron-like cells, β-catenin protein accumulation was increased in the nucleus, indicating the increased activity of Wnt/β-catenin pathway in the time-window of DA differentiation. Interestingly, DKK1 transcript level was reduced dramatically after DA induction in TM-MSCs which was accompanied by an increase in the in silico-predicted MIR9 and MIR101 levels. Measuring DKK1 expression level after overexpressing either MIR9 or MIR101 and performing luciferase assay alongside, revealed that both miR-9 and miR-101 suppress DKK1 expression and that miR-9 exerts a direct inhibitory effect on 3'UTR regulatory region. Therefore miR-9 and miR-101 might explain, at least in part, the underlying regulatory mechanism of DKK1 reduction and resulting Wnt/β-catenin pathway activation during DA neuron differentiation process.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Retinsäure, ≥98% (HPLC), powder