Merck
  • Widespread epigenetic changes to the enhancer landscape of mouse liver induced by a specific xenobiotic agonist ligand of the nuclear receptor CAR.
Widespread epigenetic changes to the enhancer landscape of mouse liver induced by a specific xenobiotic agonist ligand of the nuclear receptor CAR.
Toxicological sciences : an official journal of the Society of Toxicology (2019-06-27)
Andy Rampersaud, Nicholas J Lodato, Aram Shin, David J Waxman
ZUSAMMENFASSUNG
CAR (Nr1i3), a liver nuclear receptor and xenobiotic sensor, induces drug, steroid and lipid metabolism and dysregulates genes linked to hepatocellular carcinogenesis, but its impact on the liver epigenome is poorly understood. TCPOBOP, a halogenated xenochemical and highly specific CAR agonist ligand, induces localized chromatin opening or closing at several thousand mouse liver genomic regions, discovered as differential DNase-hypersensitive sites (ΔDHS). Active enhancer and promoter histone marks induced by TCPOBOP were enriched at opening DHS and TCPOBOP-inducible genes. Enrichment of CAR binding and CAR motifs was seen at opening DHS and their inducible drug/lipid metabolism gene targets, and at many constitutively open DHS located nearby. TCPOBOP-responsive cell cycle and DNA replication genes co-dependent on MET/EGFR signaling for induction were also enriched for CAR binding. A subset of opening DHS and many closing DHS mapping to TCPOBOP-responsive target genes did not bind CAR, indicating an indirect mechanism for their changes in chromatin accessibility. TCPOBOP-responsive DHS were also enriched for induced binding of RXRA, CEBPA and CEBPB, and for motifs for liver-enriched factors that may contribute to liver-specific transcriptional responses to TCPOBOP exposure. These studies elucidate the enhancer landscape of TCPOBOP-exposed liver and the widespread epigenetic changes that are induced by both direct and indirect mechanisms linked to CAR activation. The global maps of thousands of environmental chemical-induced epigenetic changes described here constitute a rich resource for further research on xenochemical effects on liver chromatin states and the epigenome.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
TCPOBOP, ≥98% (HPLC), solid