Merck
Streptococcus pneumoniae G5 domains bind different ligands.
Protein science : a publication of the Protein Society (2019-08-08)
Natasia Paukovich, Jasmina S Redzic, Ying-Chih Chi, Jeremy T Rahkola, Aaron Issaian, Ashley Blue, Kirk C Hansen, Edward N Janoff, Elan Z Eisenmesser
ZUSAMMENFASSUNG
Many bacterial pathogens express small G5 domains that exist in the context of various membrane-anchored proteins and these G5 domains have been associated with colonization, cellular adhesion, and biofilm formation. However, despite over a decade since the computational prediction of these G5 domains, many remain uncharacterized, particularly those from Streptococcus pneumoniae. Of five previously predicted G5 domains we found that four of these, all derived from S. pneumoniae, are independently folded modules. As one of these exhibits extreme line broadening due to self-association, we were able to use NMR solution studies to probe the potential ligand interactions of the remaining three G5 domains. None of these G5 domains engage N-acetylglucosamine (NAG) as previously predicted but do interact with other small molecules that may modulate adherence to both bacteria and host cells. Specifically, while all G5 domains tested engage Zn, only one of these G5 domains engage heparin. NMR solution structural studies of the IgA1 Protease G5 (IgA1P-G5) and endo-beta-N-acetylglucosaminidase-D G5 (ENDD-G5) also facilitated identification of the ligand binding sites and confirm the typical G5 fold that comprises two connected β-sheets with no canonical core. NMR relaxation experiments indicate flexibility on both ends and within the connecting regions between the β-sheets. Our studies thus establish a basis for future biological experiments to test whether the ligands presented here are involved in bacterial adherence, either to bacteria or to host cells.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N-Acetyl-D-Glucosamin, ≥99% (HPLC)
Sigma-Aldrich
Cholinchlorid, ≥99%
Sigma-Aldrich
Pararosanilin-Lösung -Lösung