Merck
  • Optical coherence tomography angiography to assess vascular remodeling of the choriocapillaris after low-fluence photodynamic therapy for chronic central serous chorioretinopathy.
Optical coherence tomography angiography to assess vascular remodeling of the choriocapillaris after low-fluence photodynamic therapy for chronic central serous chorioretinopathy.
Photodiagnosis and photodynamic therapy (2019-06-05)
Gilda Cennamo, Michela Cennamo, Giuseppe Caputo, Federica Mirra, Pia Clara Pafundi, Giuseppe de Crecchio, Giovanni Cennamo
ZUSAMMENFASSUNG
To evaluate the efficacy of optical coherence tomography angiography (OCTA) in identifying changes in the choriocapillaris layer after low-fluence verteporfin photodynamic therapy (vPDT) in patients affected by chronic central serous chorioretinopathy (CSCR). Low-fluence vPDT was performed on 28 eyes of 27 patients with CSCR. All patients underwent the following tests at baseline and 6 months after treatment: best corrected visual acuity (BCVA), fluorescein angiography, indocyanine green angiography, enhanced depth imaging OCT and OCTA. Subretinal fluid was completely absorbed in 18 of the 28 affected eyes (64.3%) after low-fluence vPDT ("responders"), and incompletely absorbed in 10 eyes (35.7%) ("non responders"). BCVA was significantly improved (p = 0.006) whereas central foveal thickness and choroidal foveal thickness were significantly decreased (p = 0.001 and p = 0.00 respectively) 6 months after treatment in responders. OCTA revealed a different pattern of vascular remodeling of the choriocapillaris between CSC patients who responded and those who did not respond to low-fluence vPDT.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazin, ≥99%