Merck
Anti-inflammatory and antiviral effects of minocycline in enterovirus 71 infections.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie (2019-08-05)
Yu-Ting Liao, Shih-Min Wang, Shun-Hua Chen
ZUSAMMENFASSUNG
Enterovirus 71 (EV71) brainstem encephalitis (BE) is divided into-uncomplicated BE, autonomic nervous system (ANS) dysregulation, and pulmonary edema (PE)-based on cytokine-mediated severe systemic and central nervous system (CNS) inflammatory responses. Minocycline has been found to have anti-inflammatory and immunomodulatory properties in infectious and inflammatory neurological disease models. The effects of minocycline on EV71 infection were studied in vitro and in vivo experiments. The minocycline treatment (100-300 μg/mL) on cytokine expressions and viral replications were investigated in rhabdomyosarcoma (RD), U-87MG, and THP-1 cells. The mouse-adapted-EV71 strain (MP4)-infected 7-day-old ICR mice model was used to explore the anti-inflammatory and antiviral effects of minocycline (1 and 5 μg/g) for the treatment of EV71 infection. In in vitro, minocycline reduced cytopathic effects (CPEs), viral protein expressions, viral titers, the levels of interleukin (IL)-6 and IL-8 and relative mRNA expressions of IL-12p40, IL-1β, and tumor necrosis factor (TNF) after EV71 infection. The levels of TNF, IL-1β, IL-6, and IL-8 decreased with a single dose of minocycline in EV71-infected THP-1 cells. Double-dose minocycline treatment demonstrated more effective reduction in cytokines. In the MP4-infected animal model, clinical scores, mortality rates and viral titers in various brain tissues were decreased evidently after double-dose minocycline treatment. Minocycline inhibited IL-6 and granulocyte colony-stimulating factor (G-CSF) in plasma and TNF in the cerebellum. Minocycline has properties that enable it to function both as an anti-inflammatory and antiviral agent in EV71 infection. These results evidence its potential usefulness in clinical treatment.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Minocyclin -hydrochlorid, powder