Merck
  • Chitosan/glyphosate formulation as a potential, environmental friendly herbicide with prolonged activity.
Chitosan/glyphosate formulation as a potential, environmental friendly herbicide with prolonged activity.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes (2019-08-14)
Piotr Rychter
ZUSAMMENFASSUNG
Although there are many reports on the dangers posed by glyphosate, this herbicide is still one of the most popular and widely used total weed killers. Therefore, great effort should be made to minimize exposure to this herbicide and limit its losses into the environment. The aim of this study was to prepare a new formulation consisted of various molecular weight chitosans with glyphosate and their evaluation toward active substance release, phytotoxicity, and preliminary herbicidal efficiency. The phytotoxicity study of the obtained chitosan/glyphosate formulations was determined based on the Organisation for Economic Co-operation and Development 208 guideline. Among all tested formulations evaluated for phytotoxicity, that containing the highest molecular weight of chitosan was found to be the least toxic, showing simultaneously the most effective herbicidal activity against selected weeds. The release rate of glyphosate from the obtained formulations was dependent on the molecular weight and viscosity of chitosan.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N-(Phosphonomethyl)glycin, 96%