Merck
Air-liquid interface culture changes surface properties of A549 cells.
Toxicology in vitro : an international journal published in association with BIBRA (2019-06-25)
Kristin Öhlinger, Tatjana Kolesnik, Claudia Meindl, Birgit Gallé, Markus Absenger-Novak, Dagmar Kolb-Lenz, Eleonore Fröhlich
ZUSAMMENFASSUNG
A549 cells are common models in the assessment of respiratory cytotoxicity. To provide physiologically more representative exposure conditions and increase the differentiation state, respiratory cells, for instance Calu-3 bronchial epithelial cells, are cultured at an air-liquid interface (ALI). There are indications that A549 cells also change their phenotype upon culture in ALI. The influence of culture in two variations of transwell cultures compared to conventional culture in plastic wells on the phenotype of A549 cells was studied. Cells were characterized by morphology, proliferation and transepithelial electrical resistance, whole genome transcription analysis, Western blot and immunocytochemical detection of pro-surfactant proteins. Furthermore, lipid staining, surface morphology, cell elasticity, surface tension and reaction to quartz particles were performed. Relatively small changes were noted in the expression of differentiation markers for alveolar cells but A549 cells cultured in ALI showed marked differences in lipid staining and surface morphology, surface tension and cytotoxicity of quartz particles. Data show that changes in physiological reactions of A549 cells in ALI culture were rather caused by change of surface properties than by increased expression of surfactant proteins.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Proteasehemmer-Cocktail, for use with mammalian cell and tissue extracts, DMSO solution