Merck
Loss of flow responsive Tie1 results in Impaired
Aortic valve remodeling.
Developmental biology (2019-07-19)
Xianghu Qu, Kate Violette, M K Sewell-Loftin, Jonathan Soslow, LeShana Saint-Jean, Robert B Hinton, W David Merryman, H Scott Baldwin
ZUSAMMENFASSUNG
The mechanisms regulating endothelial cell response to hemodynamic forces required for heart valve development, especially valve remodeling, remain elusive. Tie1, an endothelial specific receptor tyrosine kinase, is up-regulated by oscillating shear stress and is required for lymphatic valve development. In this study, we demonstrate that valvular endothelial Tie1 is differentially expressed in a dynamic pattern predicted by disturbed flow during valve remodeling. Following valvular endocardial specific deletion of Tie1 in mice, we observed enlarged aortic valve leaflets, decreased valve stiffness and valvular insufficiency. Valve abnormalities were only detected in late gestation and early postnatal mutant animals and worsened with age. The mutant mice developed perturbed extracellular matrix (ECM) deposition and remodeling characterized by increased glycosaminoglycan and decreased collagen content, as well as increased valve interstitial cell expression of Sox9, a transcription factor essential for normal ECM maturation during heart valve development. This study provides the first evidence that Tie1 is involved in modulation of late valve remodeling and suggests that an important Tie1-Sox9 signaling axis exists through which disturbed flows are converted by endocardial cells to paracrine Sox9 signals to modulate normal matrix remodeling of the aortic valve.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler Anti-Aktin, α-Glattmuskel - Cy3 Maus-Antikörper, clone 1A4, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Phospho-Histon H3 (Ser10)-Antikörper, Mitosemarker, Upstate®, from rabbit
Sigma-Aldrich
Hyaluronsäure-bindendes Protein, Rindernasenknorpel, biotinyliert
Sigma-Aldrich
ApopTag Plus In-situ-Apoptose-Fluoreszeinnachweis-Kit, The ApopTag Plus Fluorescein In Situ Apoptosis Detection Kit detects apoptotic cells in situ by the indirect TUNEL method, utilizing an anti-digoxigenin antibody that is conjugated to a fluorescein reporter molecule.
Sigma-Aldrich
Anti-Versican-Antikörper, a.a. 1360-1439 Maus-Versican, Chemicon®, from rabbit
Sigma-Aldrich
Phenyl-β-D-Galaktopyranosid, ≥98% (TLC)