Merck
Human FcRn Tissue Expression Profile and Half-Life in PBMCs.
Biomolecules (2019-08-25)
Yao-Yun Fan, Vahid Farrokhi, Teresa Caiazzo, Mengmeng Wang, Denise M O'Hara, Hendrik Neubert
ZUSAMMENFASSUNG
System-wide quantitative characterization of human neonatal Fc receptor (FcRn) properties is critical for understanding and predicting human PK (pharmacokinetics) as well as the distribution of mAbs and Fc-fusion proteins using PBPK (physiologically-based pharmacokinetic) modeling. To this end, tissue-specific FcRn expression and half-life are important model inputs. Herein, human FcRn tissue expression was measured by peptide immunoaffinity chromatography coupled with high-resolution mass spectrometry. FcRn concentrations across 14 human tissues ranged from low to 230 pmol per gram of tissue. Furthermore, the FcRn half-life was determined to be 11.1 h from a human stable isotope labelled leucine pulse labeling experiment. The spatial and temporal quantitative human FcRn data now promise to enable a refined PBPK model with improved accuracy of human PK predictions for Fc-containing biotherapeutics.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Trypsin aus Rinderpankreas, TPCK Treated, essentially salt-free, lyophilized powder, ≥10,000 BAEE units/mg protein