Merck
Control of bone formation by the serpentine receptor Frizzled-9.
The Journal of cell biology (2011-03-16)
Joachim Albers, Jochen Schulze, F Timo Beil, Matthias Gebauer, Anke Baranowsky, Johannes Keller, Robert P Marshall, Kristofer Wintges, Felix W Friedrich, Matthias Priemel, Arndt F Schilling, Johannes M Rueger, Kerstin Cornils, Boris Fehse, Thomas Streichert, Guido Sauter, Franz Jakob, Karl L Insogna, Barbara Pober, Klaus-Peter Knobeloch, Uta Francke, Michael Amling, Thorsten Schinke
ZUSAMMENFASSUNG
Although Wnt signaling in osteoblasts is of critical importance for the regulation of bone remodeling, it is not yet known which specific Wnt receptors of the Frizzled family are functionally relevant in this process. In this paper, we show that Fzd9 is induced upon osteoblast differentiation and that Fzd9(-/-) mice display low bone mass caused by impaired bone formation. Our analysis of Fzd9(-/-) primary osteoblasts demonstrated defects in matrix mineralization in spite of normal expression of established differentiation markers. In contrast, we observed a reduced expression of chemokines and interferon-regulated genes in Fzd9(-/-) osteoblasts. We also identified the ubiquitin-like modifier Isg15 as one potential downstream mediator of Fzd9 in these cells. Importantly, our molecular analysis further revealed that canonical Wnt signaling is not impaired in the absence of Fzd9, thus explaining the absence of a bone resorption phenotype. Collectively, our results reveal a previously unknown function of Fzd9 in osteoblasts, a finding that may have therapeutic implications for bone loss disorders.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-PTH/PTHrP-Rezeptor-Antikörper, Klon 3D1.1, clone 3D1.1, Upstate®, from mouse