Merck
Transcriptomic analysis reveals new hippocampal gene networks induced by prolactin.
Scientific reports (2019-09-26)
Erika Alejandra Cabrera-Reyes, América Vanoye-Carlo, Mauricio Rodríguez-Dorantes, Edgar Ricardo Vázquez-Martínez, Nadia Alejandra Rivero-Segura, Omar Collazo-Navarrete, Marco Cerbón
ZUSAMMENFASSUNG
Prolactin (Prl) is a pleiotropic hormone with multiple functions in several tissues and organs, including the brain. In the hippocampus, Prl has been implicated in several functions, including neuroprotection against excitotoxicity in lactating rats and in Prl-treated ovariectomized animals. However, the molecular mechanisms involved in Prl actions in the hippocampus have not been completely elucidated. The aim of this study was to analyse the hippocampal transcriptome of female Prl-treated ovariectomized rats. Transcriptomic analysis by RNASeq revealed 162 differentially expressed genes throughout 24 h of Prl treatment. Gene Ontology analysis of those genes showed that 37.65% were involved in brain processes that are regulated by the hippocampus, such as learning, memory and behaviour, as well as new processes that we did not foresee, such as glial differentiation, axogenesis, synaptic transmission, postsynaptic potential, and neuronal and glial migration. Immunodetection analysis demonstrated that Prl significantly modified microglial morphology, reduced the expression of Cd11b/c protein, and altered the content and location of the neuronal proteins Tau, Map2 and Syp, which are involved in axogenic and synaptic functions. This novel delineation of Prl activity in the hippocampus highlights its importance as a neuroactive hormone, opens a new avenue for understanding its actions and supports its participation in neuronal plasticity of this brain area.