Merck
  • Parvalbumin interneurons and calretinin fibers arising from the thalamic nucleus reuniens degenerate in the subiculum after kainic acid-induced seizures.
Parvalbumin interneurons and calretinin fibers arising from the thalamic nucleus reuniens degenerate in the subiculum after kainic acid-induced seizures.
Neuroscience (2011-05-28)
M Drexel, A P Preidt, E Kirchmair, G Sperk
ZUSAMMENFASSUNG
The subiculum is the major output area of the hippocampus. It is closely interconnected with the entorhinal cortex and other parahippocampal areas. In animal models of temporal lobe epilepsy (TLE) and in TLE patients it exerts increased network excitability and may crucially contribute to the propagation of limbic seizures. Using immunohistochemistry and in situ-hybridization we now investigated neuropathological changes affecting parvalbumin and calretinin containing neurons in the subiculum and other parahippocampal areas after kainic acid-induced status epilepticus. We observed prominent losses in parvalbumin containing interneurons in the subiculum and entorhinal cortex, and in the principal cell layers of the pre- and parasubiculum. Degeneration of parvalbumin-positive neurons was associated with significant precipitation of parvalbumin-immunoreactive debris 24 h after kainic acid injection. In the subiculum the superficial portion of the pyramidal cell layer was more severely affected than its deep part. In the entorhinal cortex, the deep layers were more severely affected than the superficial ones. The decrease in number of parvalbumin-positive neurons in the subiculum and entorhinal cortex correlated with the number of spontaneous seizures subsequently experienced by the rats. The loss of parvalbumin neurons thus may contribute to the development of spontaneous seizures. On the other hand, surviving parvalbumin neurons revealed markedly increased expression of parvalbumin mRNA notably in the pyramidal cell layer of the subiculum and in all layers of the entorhinal cortex. This indicates increased activity of these neurons aiming to compensate for the partial loss of this functionally important neuron population. Furthermore, calretinin-positive fibers terminating in the molecular layer of the subiculum, in sector CA1 of the hippocampus proper and in the entorhinal cortex degenerated together with their presumed perikarya in the thalamic nucleus reuniens. In addition, a significant loss of calretinin containing interneurons was observed in the subiculum. Notably, the loss in parvalbumin positive neurons in the subiculum equaled that in human TLE. It may result in marked impairment of feed-forward inhibition of the temporo-ammonic pathway and may significantly contribute to epileptogenesis. Similarly, the loss of calretinin-positive fiber tracts originating from the nucleus reuniens thalami significantly contributes to the rearrangement of neuronal circuitries in the subiculum and entorhinal cortex during epileptogenesis.