Merck
  • Synthesis of chitosan-grafted-poly(N-vinylcaprolactam) coated on the thiolated gold nanoparticles surface for controlled release of cisplatin.
Synthesis of chitosan-grafted-poly(N-vinylcaprolactam) coated on the thiolated gold nanoparticles surface for controlled release of cisplatin.
Carbohydrate polymers (2019-10-09)
Solmaz Banihashem, Mahshid Nikpour Nezhati, Homayon Ahmad Panahi
ZUSAMMENFASSUNG
The gold nanoparticles surface was modified by thioglycolic acid ligand and their surface was coated by the chitosan-grafted-poly(N-vinylcaprolactam) (chitosan-g-PNVCL) copolymer. The cisplatin anticancer drug was loaded into the synthesized nanocarriers and its performance was investigated for the treatment of MCF-7 breast cancer cells in vitro. The synthesized nanoparticles were characterized using FTIR, DLS, TEM, SEM, EDX and TGA analysis. The lower critical solution temperature (LCST) of PNVCL/chitosan and PNVCL/chitosan coated gold nanoparticles were found to be 38 and 39 °C, respectively. The cisplatin loading efficiency, cisplatin release from nanoparticles at different temperatures and pH values as well as the pharmacokinetic studies were examined. The maximum cisplatin release from nanoparticles was achieved at T > LCST (42 °C) and pH of 5. The Korsemeyer-Peppas model was best described the cisplatin release from nanoparticles. The maximum MCF cell death was found to be 92% using cisplatin loaded-gold/TGA/chitosan-g-PNVCL nanoparticles under an induction heating system.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N-Vinylcaprolactam, 98%, stabilized