Merck
  • Synergism of Adipocytokine Profile and ADIPOQ/TNF-α Polymorphisms in NAFLD-associated MetS Predict Colorectal Liver Metastases Outgrowth.
Synergism of Adipocytokine Profile and ADIPOQ/TNF-α Polymorphisms in NAFLD-associated MetS Predict Colorectal Liver Metastases Outgrowth.
Cancer genomics & proteomics (2019-10-30)
Rosa Divella, Antonella Daniele, Raffaele DE Luca, Antonio Mazzocca, Eustachio Ruggieri, Eufemia Savino, Porzia Casamassima, Michele Simone, Carlo Sabba, Angelo Paradiso
ZUSAMMENFASSUNG
The aim of this study was to evaluate whether the altered profile of adipocytokine and genetic fingerprint in NAFLD-associated metabolic syndrome "cluster" represents synergistic risk factors predicting onset of liver colorectal cancer metastases. A total of 165 colorectal cancer patients were enrolled, 56,3% were with metabolic syndrome/NAFLD. Serum samples were assayed for ADIPOQ, leptin and TNF-a levels by ELISA. ADIPOQ rs266729 C/G and TNF-308 A/G genotypes were analyzed in DNA isolated from whole blood. Reduction in adiponectin levels and increase in leptin and TNF-α was shown in patients with liver metastases. This trend was influenced by BMI, MetS/NAFLD, and insulin resistance. ADIPOQ G rs266729 and TNF- 308 A allele are associated with obesity, MetS/NAFLD and insulin resistance. ADIPOQ CG/GG and GA/AA TNF-alpha genotypes confer susceptibility to liver metastases. Obesity and hepatic steatosis significantly favor the development of colorectal cancer liver metastases and the individual adipocytokines genetic profile may play an important predictive role.