Merck
  • Purification, Characterization and Evaluation of Inhibitory Mechanism of ACE Inhibitory Peptides from Pearl Oyster (Pinctada fucata martensii) Meat Protein Hydrolysate.
Purification, Characterization and Evaluation of Inhibitory Mechanism of ACE Inhibitory Peptides from Pearl Oyster (Pinctada fucata martensii) Meat Protein Hydrolysate.
Marine drugs (2019-08-11)
Pengru Liu, Xiongdiao Lan, Muhammad Yaseen, Shanguang Wu, Xuezhen Feng, Liqin Zhou, Jianhua Sun, Anping Liao, Dankui Liao, Lixia Sun
ZUSAMMENFASSUNG
Angiotensin-I-converting enzyme (ACE) inhibitory peptides derived from natural products have shown a blood pressure lowering effect with no side effects. In this study, two novel ACE inhibitory peptides (His-Leu-His-Thr, HLHT and Gly-Trp-Ala, GWA) were purified from pearl oyster (Pinctada fucata martensii) meat protein hydrolysate with alkaline protease by ultrafiltration, polyethylene glycol methyl ether modified immobilized metal ion affinity medium, and reverse-phase high performance liquid chromatography. Both peptides exhibited high ACE inhibitory activity with IC50 values of 458.06 ± 3.24 μM and 109.25 ± 1.45 μM, respectively. Based on the results of a Lineweaver-Burk plot, HLHT and GWA were found to be non-competitive inhibitor and competitive inhibitor respectively, which were confirmed by molecular docking. Furthermore, the pearl oyster meat protein hydrolysate exhibited an effective antihypertensive effect on SD rats. These results conclude that pearl oyster meat protein is a potential resource of ACE inhibitory peptides and the purified peptides, HLHT and GWA, can be exploited as functional food ingredients against hypertension.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N-Hexanoyl-L-homoserin-lacton, ≥96% (HPLC)