Merck
  • A large pool of actively cycling progenitors orchestrates self-renewal and injury repair of an ectodermal appendage.
A large pool of actively cycling progenitors orchestrates self-renewal and injury repair of an ectodermal appendage.
Nature cell biology (2019-09-05)
Amnon Sharir, Pauline Marangoni, Rapolas Zilionis, Mian Wan, Tomas Wald, Jimmy K Hu, Kyogo Kawaguchi, David Castillo-Azofeifa, Leo Epstein, Kyle Harrington, Pierfrancesco Pagella, Thimios Mitsiadis, Christian W Siebel, Allon M Klein, Ophir D Klein
ZUSAMMENFASSUNG
The classical model of tissue renewal posits that small numbers of quiescent stem cells (SCs) give rise to proliferating transit-amplifying cells before terminal differentiation. However, many organs house pools of SCs with proliferative and differentiation potentials that diverge from this template. Resolving SC identity and organization is therefore central to understanding tissue renewal. Here, using a combination of single-cell RNA sequencing (scRNA-seq), mouse genetics and tissue injury approaches, we uncover cellular hierarchies and mechanisms that underlie the maintenance and repair of the continuously growing mouse incisor. Our results reveal that, during homeostasis, a group of actively cycling epithelial progenitors generates enamel-producing ameloblasts and adjacent layers of non-ameloblast cells. After injury, tissue repair was achieved through transient increases in progenitor-cell proliferation and through direct conversion of Notch1-expressing cells to ameloblasts. We elucidate epithelial SC identity, position and function, providing a mechanistic basis for the homeostasis and repair of a fast-turnover ectodermal appendage.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
(Z)-4-Hydroxytamoxifen, ≥98% Z isomer
Roche
In Situ Cell Death Detection Kit, TMR rot, sufficient for ≤50 tests