Merck
Physicochemical properties of dodecenyl succinic anhydride (DDSA) modified quinoa starch.
Food chemistry (2019-07-30)
Guantian Li, Xu Xu, Fan Zhu
ZUSAMMENFASSUNG
Quinoa starch granules were esterified with dodecenyl succinic anhydride (DDSA) to various degrees of substitution (DS) (0.0023-0.0095). Physicochemical properties and emulsification capacity of the modified starch were studied. Increasing DS increased the particle size, water solubility, and swelling power, while decreasing the gelatinization enthalpy change and relative crystallinity of the starch. The DDSA groups were suggested to be mostly located in the amorphous region of starch granules. With increasing DS, the viscosity and storage modulus (G') obtained from rheological analysis increased first and then decreased. The DDSA modified quinoa starch efficiently stabilized O/W Pickering emulsions and has some technical advantages compared to octenyl succinic anhydride (OSA)-modified quinoa starch. Increasing DS led to decreased droplet size of the emulsions and a higher capacity in stabilizing emulsions during storage for a period of 10 days. This study suggested the potential application of DDSA modified quinoa starch as an emulsifier in stabilizing Pickering emulsions.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dodecenylbernsteinsäureanhydrid, Isomermischung, technical grade, 90%