Merck
A Quantitative Multivariate Model of Human Dendritic Cell-T Helper Cell Communication.
Cell (2019-10-05)
Maximilien Grandclaudon, Marie Perrot-Dockès, Coline Trichot, Léa Karpf, Omar Abouzid, Camille Chauvin, Philémon Sirven, Wassim Abou-Jaoudé, Frédérique Berger, Philippe Hupé, Denis Thieffry, Laure Sansonnet, Julien Chiquet, Céline Lévy-Leduc, Vassili Soumelis
ZUSAMMENFASSUNG
Cell-cell communication involves a large number of molecular signals that function as words of a complex language whose grammar remains mostly unknown. Here, we describe an integrative approach involving (1) protein-level measurement of multiple communication signals coupled to output responses in receiving cells and (2) mathematical modeling to uncover input-output relationships and interactions between signals. Using human dendritic cell (DC)-T helper (Th) cell communication as a model, we measured 36 DC-derived signals and 17 Th cytokines broadly covering Th diversity in 428 observations. We developed a data-driven, computationally validated model capturing 56 already described and 290 potentially novel mechanisms of Th cell specification. By predicting context-dependent behaviors, we demonstrate a new function for IL-12p70 as an inducer of Th17 in an IL-1 signaling context. This work provides a unique resource to decipher the complex combinatorial rules governing DC-Th cell communication and guide their manipulation for vaccine design and immunotherapies.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Phorbol-12-myristat-13-acetat, ≥99% (TLC), film or powder
Sigma-Aldrich
Prostaglandin E2, synthetic, powder, BioReagent, suitable for cell culture