Merck
  • Mechanism of the allosteric activation of the ClpP protease machinery by substrates and active-site inhibitors.
Mechanism of the allosteric activation of the ClpP protease machinery by substrates and active-site inhibitors.
Science advances (2019-09-14)
Jan Felix, Katharina Weinhäupl, Christophe Chipot, François Dehez, Audrey Hessel, Diego F Gauto, Cecile Morlot, Olga Abian, Irina Gutsche, Adrian Velazquez-Campoy, Paul Schanda, Hugo Fraga
ZUSAMMENFASSUNG
Coordinated conformational transitions in oligomeric enzymatic complexes modulate function in response to substrates and play a crucial role in enzyme inhibition and activation. Caseinolytic protease (ClpP) is a tetradecameric complex, which has emerged as a drug target against multiple pathogenic bacteria. Activation of different ClpPs by inhibitors has been independently reported from drug development efforts, but no rationale for inhibitor-induced activation has been hitherto proposed. Using an integrated approach that includes x-ray crystallography, solid- and solution-state nuclear magnetic resonance, molecular dynamics simulations, and isothermal titration calorimetry, we show that the proteasome inhibitor bortezomib binds to the ClpP active-site serine, mimicking a peptide substrate, and induces a concerted allosteric activation of the complex. The bortezomib-activated conformation also exhibits a higher affinity for its cognate unfoldase ClpX. We propose a universal allosteric mechanism, where substrate binding to a single subunit locks ClpP into an active conformation optimized for chaperone association and protein processive degradation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Casein fluorescein isothiocyanate from bovine milk, Type III, essentially salt-free, lyophilized powder
Sigma-Aldrich
Allylmethylsulfon, 96%