Merck
  • Checkpoint Blockade in Combination With Doxorubicin Augments Tumor Cell Apoptosis in Osteosarcoma.
Checkpoint Blockade in Combination With Doxorubicin Augments Tumor Cell Apoptosis in Osteosarcoma.
Journal of immunotherapy (Hagerstown, Md. : 1997) (2019-06-21)
Jizhuang Wang, Chuanzhen Hu, Jun Wang, Yuhui Shen, Qiyuan Bao, Fangzhou He, Hongyi Wang, Liangzhi Gong, Zhuochao Liu, Fangqiong Hu, Jing Liang, Qi Zhou, Li Wei, Junxiang Wen, Weibin Zhang
ZUSAMMENFASSUNG
The aim of this study was to provide a basis for the theory that the combination of conventional chemotherapy and immunotherapy would be an effective treatment for osteosarcoma. Here, the expression of programmed death ligand 1 (PD-L1) in 26 clinical osteosarcoma tissue samples collected before and after chemotherapy was analyzed. The effects of osteosarcoma cells treated with doxorubicin, a conventional chemotherapeutic agent, on the proliferation and apoptosis of CD8 T lymphocytes were investigated in vitro. Thereafter, the effectiveness of doxorubicin combined with an anti-PD-L1 antibody as an osteosarcoma therapy was tested in 24 subcutaneous tumor mouse models. The results showed that the expression of PD-L1 was upregulated by chemotherapy in both the clinical osteosarcoma tissue samples and the osteosarcoma cell lines. The proliferation of CD8 T lymphocytes was inhibited, and apoptosis in CD8 T lymphocytes was enhanced by the doxorubicin-pretreated osteosarcoma cells, whereas this effect was reversed by the anti-PD-L1 antibody. A more effective result was observed when doxorubicin was combined with the anti-PD-L1 antibody in vivo. In short, the combination of conventional chemotherapy and an anti-PD-L1 antibody might be an effective option for osteosarcoma treatment, as anti-PD-L1 antibody can reverse the immunosuppression induced by chemotherapy.