Merck
  • DNA Binding and Phosphorylation Regulate the Core Structure of the NF-kappaB p50 Transcription Factor
DNA Binding and Phosphorylation Regulate the Core Structure of the NF-kappaB p50 Transcription Factor
Journal of the American Society For Mass Spectrometry (2019)
Vonderach M, et al.