Merck
  • Identification of heat shock protein 5, calnexin and integral membrane protein 2B as Adam7-interacting membrane proteins in mouse sperm.
Identification of heat shock protein 5, calnexin and integral membrane protein 2B as Adam7-interacting membrane proteins in mouse sperm.
Journal of cellular physiology (2010-10-15)
Cecil Han, Inju Park, Boyeon Lee, Sora Jin, Heejin Choi, Jun Tae Kwon, Yong-il Kwon, Do Han Kim, Zee Yong Park, Chunghee Cho
ZUSAMMENFASSUNG
In mammals, sperm acquire their motility and ability to fertilize eggs in the epididymis. This maturation process involves the acquisition of particular proteins from the epididymis. One such secretory protein specifically expressed in the epididymis is Adam7 (a disintegrin and metalloprotease 7). Previous studies have shown that Adam7 that resides in an intracellular compartment of epididymal cells is transferred to sperm membranes, where its levels are dependent on the expression of Adam2 and Adam3, which have critical roles in fertilization. Here, using a proteomics approach based on mass spectrometry, we identified proteins that interact with Adam7 in sperm membranes. This analysis revealed that Adam7 forms complexes with calnexin (Canx), heat shock protein 5 (Hspa5), and integral membrane protein 2B (Itm2b). Canx and Hspa5 are molecular chaperones, and Itm2b is a type II integral membrane protein implicated in neurodegeneration. The interaction of Adam7 with these proteins was confirmed by immunoprecipitation-Western blot analysis. We found that Adam7 and Itm2b are located in detergent-resistant regions known to be highly correlated with membrane lipid rafts. We further found that the association of Adam7 with Itm2b is remarkably promoted during sperm capacitation owing to a conformational change of Adam7 that occurs in concert with the capacitation process. Thus, our results suggest that Adam7 functions in fertilization through the formation of a chaperone complex and enhanced association with Itm2b during capacitation in sperm.