Merck
  • Ubiquitin-conjugating enzyme Ubc13 is a critical component of TNF receptor-associated factor (TRAF)-mediated inflammatory responses.
Ubiquitin-conjugating enzyme Ubc13 is a critical component of TNF receptor-associated factor (TRAF)-mediated inflammatory responses.
Proceedings of the National Academy of Sciences of the United States of America (2007-04-04)
Toru Fukushima, Shu-ichi Matsuzawa, Christina L Kress, Jean Marie Bruey, Maryla Krajewska, Sophie Lefebvre, Juan M Zapata, Ze'ev Ronai, John C Reed
ZUSAMMENFASSUNG
Ubc13 is a ubiquitin-conjugating enzyme responsible for noncanonical ubiquitination of TNF receptor-associated factor (TRAF)-family adapter proteins involved in Toll-like receptor and TNF-family cytokine receptor signaling, which are regulators of innate immunity. Gene ablation was used to study the function of Ubc13 in mice. Whereas homozygous ubc13 gene disruption resulted in embryonic lethality, heterozygous ubc13(+/-) mice appeared normal, without alterations in immune cell populations. Haploinsufficient ubc13(+/-) mice were resistant to lipopolysaccharide-induced lethality, and demonstrated reduced in vivo ubiquitination of TRAF6. Macrophages and splenocytes isolated from ubc13(+/-) mice exhibited reduced lipopolysaccharide-inducible cytokine secretion and impaired activation of TRAF-dependent signal transduction pathways (NF-kappaB, JNK, and p38 MAPK). These findings document a critical role for Ubc13 in inflammatory responses and suggest that agents reducing Ubc13 activity could have therapeutic utility.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Lipopolysaccharide aus Escherichia coli O26:B6, ≥10,000 EU/mg, purified by phenol extraction