Merck
  • Correlation of mutant menin stability with clinical expression of multiple endocrine neoplasia type 1 and its incomplete forms.
Correlation of mutant menin stability with clinical expression of multiple endocrine neoplasia type 1 and its incomplete forms.
Cancer science (2011-08-09)
Satoko Shimazu, Yuko Nagamura, Hiroko Yaguchi, Naganari Ohkura, Toshihiko Tsukada
ZUSAMMENFASSUNG
Germline mutations of the tumor suppressor gene MEN1 are found not only in typical multiple endocrine neoplasia type 1 (MEN1) but also in its incomplete forms such as familial isolated hyperparathyroidism (FIHP) and apparently sporadic parathyroid tumor (ASPT). No definitive genotype-phenotype correlation has been established between these clinical forms and MEN1 gene mutations. We previously demonstrated that mutant menin proteins associated with MEN1 are rapidly degraded by the ubiquitin-proteasome pathway. To examine whether the intracellular stability of mutant menin is correlated with clinical phenotypes, we developed a method of evaluating menin stability and examined 20 mutants associated with typical MEN1 (17 missense, two in-frame deletion, one nonsense) and 21 mutants associated with FIHP or ASPT (19 missense, two in-frame deletion). All tested mutants associated with typical MEN1 showed reduced stability. Some missense and in-frame deletion mutants (G28A, R171W, T197I, E255K, E274A, Y353del and E366D) associated with FIHP or ASPT were almost as stable as or only slightly less stable than wild-type menin, while others were as unstable as those associated with typical MEN1. Some stable mutants exhibited substantial biological activities when tested by JunD-dependent transactivation assay. These findings suggest that certain missense and in-frame mutations are fairly stable and retain intrinsic biological activity, and might be specifically associated with incomplete clinical phenotypes. The menin stability test will provide useful information for the management of patients carrying germline MEN1 mutations especially when they have missense or in-frame variants of ambiguous clinical significance.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonales ANTI-FLAG®-M2-FITC in Maus hergestellte Antikörper, clone M2, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoklonale ANTI-FLAG® M2-Alkalische Phosphatase in Maus hergestellte Antikörper, clone M2, purified immunoglobulin, buffered aqueous glycerol solution