Merck
A Role for H2O2 and TRPM2 in the Induction of Cell Death: Studies in KGN Cells.
Antioxidants (Basel, Switzerland) (2019-11-02)
Carsten Theo Hack, Theresa Buck, Konstantin Bagnjuk, Katja Eubler, Lars Kunz, Doris Mayr, Artur Mayerhofer
ZUSAMMENFASSUNG
Recent studies showed that KGN cells, derived from a human granulosa cell tumor (GCT), express NADPH oxidase 4 (NOX4), an important source of H2O2. Transient receptor potential melastatin 2 (TRPM2) channel is a Ca2+ permeable cation channel that can be activated by H2O2 and plays an important role in cellular functions. It is also able to promote susceptibility to cell death. We studied expression and functionality of TRPM2 in KGN cells and examined GCT tissue microarrays (TMAs) to explore in vivo relevance. We employed live cell, calcium and mitochondrial imaging, viability assays, fluorescence activated cell sorting (FACS) analysis, Western blotting and immunohistochemistry. We confirmed that KGN cells produce H2O2 and found that they express functional TRPM2. H2O2 increased intracellular Ca2+ levels and N-(p-Amylcinnamoyl)anthranilic acid (ACA), a TRPM2 inhibitor, blocked this action. H2O2 caused mitochondrial fragmentation and apoptotic cell death, which could be attenuated by a scavenger (Trolox). Immunohistochemistry showed parallel expression of NOX4 and TRPM2 in all 73 tumor samples examined. The results suggest that GCTs can be endowed with a system that may convey susceptibility to cell death. If so, induction of oxidative stress may be beneficial in GCT therapy. Our results also imply a therapeutic potential for TRPM2 as a drug target in GCTs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonales Anti-β-Aktin in Maus hergestellte Antikörper, clone AC-15, ascites fluid
Sigma-Aldrich
Ampliflu Rot, suitable for fluorescence, ≥98.0% (HPLC)
Sigma-Aldrich
Kartogenin, ≥98% (HPLC)
Sigma-Aldrich
Anti-TRPM2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution