Merck
  • Induction of 3-hydroxy-3-methylglutaryl coenzyme A reductase activity in human fibroblasts incubated with compactin (ML-236B), a competitive inhibitor of the reductase.
Induction of 3-hydroxy-3-methylglutaryl coenzyme A reductase activity in human fibroblasts incubated with compactin (ML-236B), a competitive inhibitor of the reductase.
The Journal of biological chemistry (1978-02-25)
M S Brown, J R Faust, J L Goldstein, I Kaneko, A Endo
PMID624722
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Mevastatin, ≥98% (HPLC), powder or crystals