Merck
mGluR6 deletion renders the TRPM1 channel in retina inactive.
Journal of neurophysiology (2011-12-02)
Ying Xu, Anuradha Dhingra, Marie E Fina, Chieko Koike, Takahisa Furukawa, Noga Vardi
ZUSAMMENFASSUNG
In darkness, glutamate released from photoreceptors activates the metabotropic glutamate receptor 6 (mGluR6) on retinal ON bipolar cells. This activates the G protein G(o), which then closes transient receptor potential melastatin 1 (TRPM1) channels, leading to cells' hyperpolarization. It has been generally assumed that deleting mGluR6 would render the cascade inactive and the ON bipolar cells constitutively depolarized. Here we show that the rod bipolar cells in mGluR6-null mice were hyperpolarized. The slope conductance of the current-voltage curves and the current noise were smaller than in wild type. Furthermore, while in wild-type rod bipolar cells, TRPM1 could be activated by local application of capsaicin; in null cells, it did not. These results suggest that the TRPM1 channel in mGluR6-null rod bipolar cells is inactive. To explore the reason for this lack of activity, we tested if mGluR6 deletion affected expression of cascade components. Immunostaining for G protein subunit candidates Gα(o), Gβ(3), and Gγ(13) showed no significant changes in their expression or distribution. Immunostaining for TRPM1 in the dendritic tips was greatly reduced, but the channel was still present in the soma and primary dendrites of mGluR6-null bipolar cells, where a certain fraction of TRPM1 appears to localize to the plasma membrane. Consequently, the lack of TRPM1 activity in the null retina is unlikely to be due to failure of the channels to localize to the plasma membrane. We speculate that, to be constitutively active, TRPM1 channels in ON bipolar cells have to be in a complex, or perhaps require an unidentified factor.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Cystein, from non-animal source, BioReagent, suitable for cell culture, ≥98%