Merck
  • CD133+ colon cancer cells are more interactive with the tumor microenvironment than CD133- cells.
CD133+ colon cancer cells are more interactive with the tumor microenvironment than CD133- cells.
Laboratory investigation; a journal of technical methods and pathology (2011-12-14)
Celia Chao, J Russ Carmical, Kirk L Ives, Thomas G Wood, Judith F Aronson, Guillermo A Gomez, Clarisse D Djukom, Mark R Hellmich
ZUSAMMENFASSUNG
Experimental data indicate that colorectal cancer cells with CD133 expression exhibit enhanced tumorigenicity over CD133-negative (CD133-) cells. We hypothesized that CD133-positive (CD133+) cells, compared with CD133-, are more tumorigenic because they are more interactive with and responsive to their stromal microenvironment. Freshly dissected and dissociated cells from a primary colon cancer were separated into carcinoma-associated fibroblasts (CAF) and the epithelial cells; the latter were further separated into CD133+ and CD133- cells using fluorescence-activated cell sorter. The CD133+ cells formed large tumors in non-obese diabetic-severe combined immunodeficient (NOD-SCID) mice, demonstrating the phenotypic cellular diversity of the original tumor, whereas CD133- cells were unable to sustain significant growth. Affymetrix gene array analyses using t-test, fold-change and multiple test correction identified candidate genes that were differentially expressed between the CD133+ vs CD133- cells. RT-PCR verified differences in expression for 30 of the 46 genes selected. Genes upregulated (+ vs - cells) included CD133 (9.3-fold) and CXCR4 (4-fold), integrin β8 and fibroblast growth factor receptor 2. The CAF highly express the respective ligands: stromal-derived factor-1 (SDF-1), vitronectin and FGF family members, suggesting a reciprocal relationship between the CD133+ and CAF cells. SDF-1 caused an increase in intracellular calcium in cells expressing both CD133 and CXCR4, confirming functional CXCR4. The CD133+/CXCR4+ phenotype is increased to 32% when the cells are grown in suspension compared with only 9% when the cells were allowed to attach. In Matrigel 3-D culture, the CD133+/CXCR4+ group treated with SDF-1 grew more colonies compared with vehicle, as well as significantly larger colony sizes of tumor spheres. These data demonstrate proof of principle that the enhanced tumorigenic potential of CD133+, compared with CD133-, cells is due to their increased ability to interact with their neighboring CAF.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonales Anti-Vimentin in Maus hergestellte Antikörper, clone V9, ascites fluid