Merck
  • Molecular mechanism for growth suppression of human hepatocellular carcinoma cells by acyclic retinoid.
Molecular mechanism for growth suppression of human hepatocellular carcinoma cells by acyclic retinoid.
Carcinogenesis (2003-06-17)
Rie Matsushima-Nishiwaki, Masataka Okuno, Yukihiko Takano, Soichi Kojima, Scott L Friedman, Hisataka Moriwaki
ZUSAMMENFASSUNG
We have reported previously that acyclic retinoid, a synthetic retinoid X receptor alpha (RXRalpha)-ligand, suppresses the development of hepatocellular carcinoma (HCC) in patients with chronic liver disease. On the other hand, HCCs become refractory to physiological concentrations of the natural RXRalpha-ligand, 9-cis retinoic acid (9cRA), due to extracellular signal-regulated kinase (Erk) 1/2-mediated phosphorylation and inactivation of RXRalpha. Here, we show that acyclic retinoid restores the function of RXRalpha in human HCC-derived HuH7 cells by inactivating the Ras-Erk 1/2 signaling system, thereby dephosphorylating RXRalpha. In contrast, 9cRA failed to suppress phosphoErk 1/2 levels and subsequent RXRalpha phosphorylation. Although 9cRA also suppressed Ras activity, it simultaneously down-regulated mitogen-activated protein kinase phosphatase-1, an enzyme that inactivates Erk, thereby leaving the phosphorylation status of Erk unchanged. A combination of 9cRA (a potent ligand) and acyclic retinoid (a weak ligand preventing phosphorylation) resulted in a marked cooperation in transactivation via the RXR-response element and in inhibiting the proliferation of HuH7 cells. These events provide a novel molecular basis for the antitumor activity of acyclic retinoid against HCC.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonales Anti-Phosphoserin, clone PSR-45, ascites fluid
Sigma-Aldrich
Peretinoin, ≥97% (HPLC)