Merck
Congenital Zika syndrome is associated with maternal protein malnutrition.
Science advances (2020-01-18)
J Barbeito-Andrés, P Pezzuto, L M Higa, A A Dias, J M Vasconcelos, T M P Santos, J C C G Ferreira, R O Ferreira, F F Dutra, A D Rossi, R V Barbosa, C K N Amorim, M P C De Souza, L Chimelli, R S Aguiar, P N Gonzalez, F A Lara, M C Castro, Z Molnár, R T Lopes, M T Bozza, J L S G Vianez, C G Barbeito, P Cuervo, M Bellio, A Tanuri, P P Garcez
ZUSAMMENFASSUNG
Zika virus (ZIKV) infection during pregnancy is associated with a spectrum of developmental impairments known as congenital Zika syndrome (CZS). The prevalence of this syndrome varies across ZIKV endemic regions, suggesting that its occurrence could depend on cofactors. Here, we evaluate the relevance of protein malnutrition for the emergence of CZS. Epidemiological data from the ZIKV outbreak in the Americas suggest a relationship between undernutrition and cases of microcephaly. To experimentally examine this relationship, we use immunocompetent pregnant mice, which were subjected to protein malnutrition and infected with a Brazilian ZIKV strain. We found that the combination of protein restriction and ZIKV infection leads to severe alterations of placental structure and embryonic body growth, with offspring displaying a reduction in neurogenesis and postnatal brain size. RNA-seq analysis reveals gene expression deregulation required for brain development in infected low-protein progeny. These results suggest that maternal protein malnutrition increases susceptibility to CZS.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Anti-Iba1/AIF1-Antikörper, clone 20A12.1, from mouse
Sigma-Aldrich
Anti-Ki-67-Antikörper, Chemicon®, from rabbit